Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abcb9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019875.2 | NP_063928.2 | 10 - 11 | 2130 | 74 | |
GenBank mRNA | AB045382.2 | - | 9 - 10 | 1950 | 74 | |
AB093298.1 | - | 10 - 11 | 2112 | 74 | ||
AF216495.1 | - | 10 - 11 | 2123 | 74 | ||
AK044140.1 | - | 10 - 11 | 2108 | 74 | ||
BC053014.1 | - | 10 - 11 | 2083 | 5686698 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Transporter Mammalian Gene Collection Mus musculus Probe spans exons |