Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp6v0a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011596.5 | NP_035726.2 | 243 | 104 | ||
GenBank mRNA | AB022323.1 | - | 209 | 104 | ||
AF218252.1 | - | 228 | 104 | |||
AK155055.1 | - | 220 | 104 | |||
AK170122.1 | - | 230 | 104 | |||
BC018369.1 | - | 196 | 104 | |||
BC108992.1 | - | 140 | 40047833 | 104 | ||
BC112905.1 | - | 203 | 3670722 | 104 | ||
CJ076040.1 | - | 243 | 104 | |||
M31226.1 | - | 139 | 104 | |||
X55184.1 | - | 140 | 104 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Transporter Mammalian Gene Collection Mus musculus 1700 Probe spans exons |