Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rasal1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001281999.1 | NP_001268928.1 | 9 - 10 | 1120 | 89 | |
NM_013832.4 | NP_038860.2 | 10 - 11 | 1096 | 89 | ||
GenBank mRNA | AF086714.1 | - | 9 - 10 | 968 | 89 | |
AK156050.1 | - | 9 - 10 | 1084 | 89 | ||
BC005418.1 | - | 10 - 11 | 1110 | 3488024 | 89 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |