Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Anapc7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019805.4 | NP_062779.3 | 1 - 2 | 240 | 87 | |
GenBank mRNA | AF076607.2 | - | 1 - 2 | 204 | 87 | |
AK030775.1 | - | 1 - 2 | 205 | 87 | ||
AK049968.1 | - | 1 - 2 | 178 | 87 | ||
AK137286.1 | - | 1 - 2 | 204 | 87 | ||
AK150764.1 | - | 1 - 2 | 204 | 87 | ||
AK150891.1 | - | 1 - 2 | 204 | 87 | ||
AK153114.1 | - | 1 - 2 | 204 | 87 | ||
AK159523.1 | - | 1 - 2 | 172 | 87 | ||
AK164202.1 | - | 1 - 2 | 209 | 87 | ||
BC006635.1 | - | 1 - 2 | 193 | 3482261 | 87 | |
BY128328.1 | - | 1 - 2 | 241 | 87 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |