Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atxn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009125.2 | NP_033151.2 | 19 - 20 | 3126 | 67 | |
GenBank mRNA | AF041472.1 | - | 19 - 20 | 3125 | 67 | |
AK135879.1 | - | 9 - 10 | 1287 | 67 | ||
AK135976.1 | - | 14 - 15 | 2083 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mus musculus Probe spans exons |