Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lig1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083188.1 | NP_001076657.1 | 13 - 14 | 1515 | 78 | |
NM_001199310.1 | NP_001186239.1 | 13 - 14 | 1523 | 78 | ||
NM_010715.2 | NP_034845.2 | 13 - 14 | 1440 | 78 | ||
GenBank mRNA | AK153993.1 | - | 13 - 14 | 1402 | 78 | |
AK161793.1 | - | 6 - 7 | 598 | 78 | ||
AK168643.1 | - | 13 - 14 | 1517 | 78 | ||
BC028287.1 | - | 13 - 14 | 1365 | 4911261 | 78 | |
BC138541.1 | - | 13 - 14 | 1523 | 8861561 | 78 | |
BC138542.1 | - | 13 - 14 | 1523 | 8861562 | 78 | |
U04674.1 | - | 12 - 13 | 1308 | 78 | ||
U19604.1 | - | 13 - 14 | 1553 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |