Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Csf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113529.1 | NP_001107001.1 | 1 - 2 | 443 | 112 | |
NM_001113530.1 | NP_001107002.1 | 1 - 2 | 443 | 112 | ||
NM_007778.4 | NP_031804.3 | 1 - 2 | 443 | 112 | ||
GenBank mRNA | AK136808.1 | - | 1 - 2 | 217 | 112 | |
AK138489.1 | - | 1 - 2 | 217 | 112 | ||
AK154261.1 | - | 1 - 2 | 444 | 112 | ||
AK154872.1 | - | 1 - 2 | 217 | 112 | ||
AK160995.1 | - | 1 - 2 | 217 | 112 | ||
AK166370.1 | - | 1 - 2 | 216 | 112 | ||
AK170154.1 | - | 1 - 2 | 153 | 112 | ||
BC025593.1 | - | 1 - 2 | 229 | 112 | ||
BC066187.1 | - | 1 - 2 | 197 | 30106218 | 112 | |
BC066200.1 | - | 1 - 2 | 197 | 30100524 | 112 | |
BC066205.1 | - | 1 - 2 | 197 | 30100601 | 112 | |
CA328312.1 | - | 1 - 2 | 383 | 112 | ||
M15692.1 | - | 1 - 2 | 133 | 109 | ||
M21149.1 | - | 1 - 2 | 106 | 112 | ||
M21952.1 | - | 1 - 2 | 196 | 112 | ||
S78392.1 | - | 1 - 2 | 112 | |||
X05010.1 | - | 1 - 2 | 218 | 112 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |