Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mfhas1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081279.1 | NP_001074748.1 | 1 - 2 | 3561 | 76 | |
GenBank mRNA | AK081646.1 | - | 1 - 2 | 526 | 76 | |
AK132349.1 | - | 1 - 2 | 2770 | 76 | ||
AK171731.1 | - | 1 - 2 | 2770 | 76 | ||
BC082308.1 | - | 1 - 2 | 3561 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |