Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eif3h | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_080635.1 | NP_542366.1 | 2 - 3 | 288 | 60 | |
GenBank mRNA | AK020033.1 | - | 2 - 3 | 311 | 60 | |
AK146200.1 | - | 2 - 3 | 311 | 60 | ||
AK149081.1 | - | 2 - 3 | 300 | 60 | ||
AK159113.1 | - | 2 - 3 | 301 | 60 | ||
AK160335.1 | - | 3 - 4 | 312 | 60 | ||
AK167651.1 | - | 2 - 3 | 291 | 60 | ||
AK167815.1 | - | 2 - 3 | 309 | 60 | ||
AK167844.1 | - | 2 - 3 | 311 | 60 | ||
AK167928.1 | - | 2 - 3 | 315 | 60 | ||
AK168108.1 | - | 2 - 3 | 313 | 60 | ||
AK168123.1 | - | 2 - 3 | 331 | 60 | ||
AK168259.1 | - | 2 - 3 | 311 | 60 | ||
AK168274.1 | - | 2 - 3 | 311 | 60 | ||
AK168553.1 | - | 2 - 3 | 313 | 60 | ||
AK169416.1 | - | 2 - 3 | 312 | 60 | ||
BC014755.1 | - | 2 - 3 | 288 | 4504013 | 60 | |
BC086915.1 | - | 2 - 3 | 310 | 30122747 | 60 | |
BC091728.1 | - | 2 - 3 | 296 | 2647034 | 60 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |