Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1500003762.mm.1 |
NM_001164036.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500003762.mm.1 |
NM_001164036.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500003762.mm.1 |
NM_001164037.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500003762.mm.1 |
NM_001164037.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500003762.mm.1 |
NM_001164038.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500003762.mm.1 |
NM_001164038.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500003762.mm.1 |
NM_001164039.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500003762.mm.1 |
NM_001164039.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500003762.mm.1 |
NM_001164040.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500003762.mm.1 |
NM_001164040.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500003762.mm.1 |
NM_008529.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500003762.mm.1 |
NM_008529.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500003762.mm.1 |
TC1500000549.mm.1 |
MTA TC |