Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mtmr7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001040699.1 | NP_001035789.1 | 10 - 11 | 1223 | 75 | |
GenBank mRNA | AF073882.1 | - | 3 - 4 | 282 | 75 | |
AK028160.1 | - | 10 - 11 | 1189 | 75 | ||
AK081973.1 | - | 10 - 11 | 1209 | 75 | ||
AK148768.1 | - | 10 - 11 | 1224 | 75 | ||
BC032254.1 | - | 10 - 11 | 1225 | 5368816 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Protein Phosphatase Mammalian Gene Collection Mus musculus Probe spans exons |