Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fat1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081286.2 | NP_001074755.2 | 16 - 17 | 10358 | 85 | |
GenBank mRNA | AJ250768.1 | - | 16 - 17 | 10358 | 85 | |
BC041794.1 | - | 2 - 3 | 211 | 85 | ||
BC100554.1 | - | 3 - 4 | 477 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |