Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Triml1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177742.4 | NP_808410.1 | 1 - 2 | 511 | 100 | |
GenBank mRNA | AK077092.1 | - | 1 - 2 | 513 | 100 | |
BC141202.1 | - | 1 - 2 | 490 | 9055814 | 100 | |
BC145531.1 | - | 1 - 2 | 490 | 9054094 | 100 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |