Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1500003050.mm.1 |
NM_001164604.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500003050.mm.1 |
NM_001164604.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500003050.mm.1 |
NM_026746.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500003050.mm.1 |
NM_026746.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500003050.mm.1 |
TC1500000433.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500003052.mm.1 |
NM_001164604.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500003052.mm.1 |
NM_001164604.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500003052.mm.1 |
NM_026746.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500003052.mm.1 |
NM_026746.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500003052.mm.1 |
TC1500000433.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500003054.mm.1 |
NM_001164604.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500003054.mm.1 |
NM_001164604.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500003054.mm.1 |
NM_026746.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500003054.mm.1 |
NM_026746.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500003054.mm.1 |
TC1500000433.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500003051.mm.1 |
NM_001164604.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500003051.mm.1 |
NM_001164604.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500003051.mm.1 |
NM_026746.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500003051.mm.1 |
NM_026746.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500003051.mm.1 |
TC1500000433.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500003053.mm.1 |
NM_001164604.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500003053.mm.1 |
NM_001164604.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500003053.mm.1 |
NM_026746.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500003053.mm.1 |
NM_026746.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500003053.mm.1 |
TC1500000433.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500003055.mm.1 |
NM_001164604.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500003055.mm.1 |
NM_001164604.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500003055.mm.1 |
TC1500000433.mm.1 |
MTA TC |