Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam20c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_030565.6 | NP_085042.2 | 2 - 3 | 1325 | 84 | |
GenBank mRNA | AK149780.1 | - | 3 - 4 | 1405 | 84 | |
AK154810.1 | - | 2 - 3 | 1321 | 84 | ||
AK220286.1 | - | 2 - 3 | 842 | 84 | ||
AY778962.1 | - | 2 - 3 | 1306 | 84 | ||
BC025814.1 | - | 2 - 3 | 1276 | 84 | ||
BC025826.1 | - | 2 - 3 | 1306 | 84 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |