Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp68 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044747.2 | NP_001038212.1 | 2 - 3 | 247 | 73 | |
GenBank mRNA | AK044831.1 | - | 2 - 3 | 217 | 73 | |
BY088951.1 | - | 2 - 3 | 250 | 73 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mus musculus Probe spans exons |