Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wbscr22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025375.3 | NP_079651.2 | 3 - 4 | 344 | 69 | |
GenBank mRNA | AF412035.1 | - | 2 - 3 | 116 | 69 | |
AK003386.1 | - | 2 - 3 | 117 | 69 | ||
AK011005.1 | - | 2 - 3 | 124 | 69 | ||
AK151986.1 | - | 2 - 3 | 125 | 69 | ||
BC093484.1 | - | 2 - 3 | 94 | 6735279 | 69 | |
BC117986.1 | - | 3 - 4 | 344 | 40102067 | 69 | |
BC117987.1 | - | 3 - 4 | 344 | 40102068 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |