Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fis1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163243.1 | NP_001156715.1 | 1 - 2 | 314 | 61 | |
GenBank mRNA | CA322565.1 | - | 1 - 2 | 314 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Mus musculus Probe spans exons |