Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ephb4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001159571.1 | NP_001153043.1 | 15 - 16 | 3197 | 69 | |
NM_010144.6 | NP_034274.4 | 15 - 16 | 3170 | 69 | ||
GenBank mRNA | AK046884.1 | - | 15 - 16 | 3135 | 69 | |
AK049340.1 | - | 15 - 16 | 3171 | 69 | ||
AK132392.1 | - | 15 - 16 | 3170 | 69 | ||
AK143422.1 | - | 3140 | 69 | |||
BC013342.1 | - | 15 - 16 | 2858 | 69 | ||
BC016505.1 | - | 15 - 16 | 3122 | 3709926 | 69 | |
BC090839.1 | - | 15 - 16 | 2947 | 69 | ||
U06834.1 | - | 16 - 17 | 2783 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protein Kinase Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |