Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Asl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133768.4 | NP_598529.1 | 1 - 2 | 43 | 85 | |
GenBank mRNA | AK076453.1 | - | 1 - 2 | 58 | 85 | |
AK140160.1 | - | 1 - 2 | 58 | 85 | ||
BC016670.1 | - | 1 - 2 | 43 | 2648753 | 85 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |