Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cops4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012001.2 | NP_036131.1 | 1 - 2 | 248 | 75 | |
GenBank mRNA | AF071314.1 | - | 1 - 2 | 117 | 75 | |
AK003752.1 | - | 1 - 2 | 110 | 75 | ||
AK009955.1 | - | 1 - 2 | 120 | 75 | ||
AK162763.1 | - | 1 - 2 | 114 | 75 | ||
AK167833.1 | - | 1 - 2 | 134 | 75 | ||
BC116800.1 | - | 1 - 2 | 102 | 40126119 | 75 | |
BC116826.1 | - | 1 - 2 | 102 | 40126145 | 75 | |
CX203220.1 | - | 1 - 2 | 248 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |