Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Daglb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_144915.3 | NP_659164.2 | 1 - 2 | 284 | 65 | |
GenBank mRNA | AK086747.1 | - | 1 - 2 | 194 | 65 | |
AK087884.1 | - | 1 - 2 | 139 | 65 | ||
AK135554.1 | - | 1 - 2 | 164 | 65 | ||
AK136372.1 | - | 1 - 2 | 142 | 65 | ||
BC016105.1 | - | 1 - 2 | 180 | 4921222 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |