Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arhgap24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001286468.1 | NP_001273397.1 | 1 - 2 | 150 | 61 | |
GenBank mRNA | BB843608.1 | - | 1 - 2 | 151 | 61 | |
BC023344.1 | - | 1 - 2 | 164 | 5064885 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |