Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Glmn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001161738.1 | NP_001155210.1 | 7 - 8 | 839 | 163 | |
NM_001161739.1 | NP_001155211.1 | 7 - 8 | 853 | 163 | ||
NM_133248.2 | NP_573511.2 | 7 - 8 | 853 | 163 | ||
GenBank mRNA | AJ566083.1 | - | 6 - 7 | 721 | 163 | |
AK034160.1 | - | 7 - 8 | 840 | 163 | ||
AK165661.1 | - | 7 - 8 | 824 | 163 | ||
AK169045.1 | - | 7 - 8 | 825 | 163 | ||
AK172659.1 | - | 7 - 8 | 791 | 163 | ||
BC003446.1 | - | 7 - 8 | 789 | 2651614 | 163 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |