Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0500008244.mm.1 |
TC0500000971.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500008244.mm.1 |
NM_001163035.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500008244.mm.1 |
NM_001163035.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500008244.mm.1 |
NM_026421.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500008244.mm.1 |
NM_026421.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500008244.mm.1 |
NR_027990.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500008244.mm.1 |
NR_027990.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500008242.mm.1 |
TC0500000971.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500008242.mm.1 |
NM_001163035.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500008242.mm.1 |
NM_001163035.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500008242.mm.1 |
NM_026421.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500008242.mm.1 |
NM_026421.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500008242.mm.1 |
NR_027990.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500008242.mm.1 |
NR_027990.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500008241.mm.1 |
TC0500000971.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500008241.mm.1 |
NM_001163035.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500008241.mm.1 |
NM_001163035.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500008241.mm.1 |
NM_026421.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500008241.mm.1 |
NM_026421.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500008241.mm.1 |
NR_027990.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500008241.mm.1 |
NR_027990.1 |
Refseq |