Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyth3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163548.1 | NP_001157020.1 | 4 - 5 | 771 | 74 | |
NM_011182.4 | NP_035312.3 | 4 - 5 | 447 | 74 | ||
GenBank mRNA | AB013470.1 | - | 4 - 5 | 301 | 74 | |
AF084221.1 | - | 4 - 5 | 341 | 74 | ||
AK016645.1 | - | 4 - 5 | 773 | 74 | ||
AK036191.1 | - | 4 - 5 | 361 | 74 | ||
AK142407.1 | - | 4 - 5 | 436 | 74 | ||
AK159229.1 | - | 4 - 5 | 419 | 74 | ||
AK168566.1 | - | 4 - 5 | 361 | 74 | ||
AK220565.1 | - | 4 - 5 | 427 | 74 | ||
BC007189.1 | - | 3 - 4 | 188 | 74 | ||
BC035296.1 | - | 4 - 5 | 388 | 3969080 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |