Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rac1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009007.2 | NP_033033.1 | 2 - 3 | 300 | 97 | |
GenBank mRNA | AK008435.1 | - | 2 - 3 | 301 | 97 | |
AK009017.1 | - | 2 - 3 | 301 | 97 | ||
AK011072.2 | - | 2 - 3 | 301 | 97 | ||
AK034601.1 | - | 2 - 3 | 301 | 97 | ||
AK047969.1 | - | 2 - 3 | 303 | 97 | ||
AK076023.1 | - | 2 - 3 | 342 | 97 | ||
AK088825.1 | - | 2 - 3 | 312 | 97 | ||
AK166383.1 | - | 2 - 3 | 307 | 97 | ||
AK199407.1 | - | 2 - 3 | 112 | 97 | ||
BC003828.1 | - | 2 - 3 | 301 | 3593957 | 97 | |
BC051053.1 | - | 2 - 3 | 300 | 5038182 | 97 | |
CX242948.1 | - | 2 - 3 | 337 | 97 | ||
JN971018.1 | - | 2 - 3 | 104 | 97 | ||
X57277.1 | - | 2 - 3 | 244 | 97 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Inventoried Probe spans exons |