Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nrbp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_144847.1 | NP_659096.1 | 7 - 8 | 637 | 68 | |
GenBank mRNA | BC011468.1 | - | 7 - 8 | 570 | 4012880 | 68 |
BC012437.1 | - | 7 - 8 | 637 | 3710089 | 68 | |
BC025042.1 | - | 2 - 3 | 99 | 68 | ||
BC068117.1 | - | 12 - 13 | 1285 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protein Kinase Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |