Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Znhit1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC026751.1 | - | 1 - 2 | 40 | 3153902 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |