Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cox19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_197980.1 | NP_932097.1 | 1 - 2 | 99 | 82 | |
GenBank mRNA | AK002486.1 | - | 1 - 2 | 115 | 82 | |
AK003697.1 | - | 1 - 2 | 143 | 82 | ||
AK007984.1 | - | 1 - 2 | 83 | |||
AK013260.1 | - | 1 - 2 | 115 | 82 | ||
AK013289.1 | - | 1 - 2 | 143 | 82 | ||
AK148526.1 | - | 1 - 2 | 72 | 82 | ||
BC031792.1 | - | 1 - 2 | 99 | 4193998 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |