Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Svop | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026805.1 | NP_081081.1 | 7 - 8 | 836 | 60 | |
GenBank mRNA | AK029670.1 | - | 6 - 7 | 545 | 60 | |
AK047655.1 | - | 7 - 8 | 828 | 60 | ||
AK047764.1 | - | 7 - 8 | 837 | 60 | ||
AK053833.1 | - | 7 - 8 | 827 | 60 | ||
AK082744.1 | - | 7 - 8 | 838 | 60 | ||
BC059878.1 | - | 7 - 8 | 789 | 6812082 | 60 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Transporter Mammalian Gene Collection Mus musculus Probe spans exons |