Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mlxip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133917.3 | NP_598678.2 | 4 - 5 | 828 | 72 | |
NM_177582.3 | NP_808250.1 | 4 - 5 | 828 | 72 | ||
GenBank mRNA | AK047475.1 | - | 4 - 5 | 828 | 72 | |
AY968204.1 | - | 4 - 5 | 696 | 72 | ||
BC137770.1 | - | 4 - 5 | 755 | 8860789 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |