Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mlxip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133917.3 | NP_598678.2 | 8 - 9 | 1219 | 66 | |
NM_177582.3 | NP_808250.1 | 8 - 9 | 1219 | 66 | ||
GenBank mRNA | AK047475.1 | - | 8 - 9 | 1219 | 66 | |
AY968204.1 | - | 8 - 9 | 1087 | 66 | ||
BC137770.1 | - | 8 - 9 | 1146 | 8860789 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Mus musculus Probe spans exons |