Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Usp42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029749.2 | NP_084025.2 | 2 - 3 | 365 | 100 | |
GenBank mRNA | AY336567.1 | - | 1 - 2 | 238 | 100 | |
BC137852.1 | - | 1 - 2 | 258 | 8860871 | 100 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Protease Mammalian Gene Collection Mus musculus Probe spans exons |