Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chfr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001289577.1 | NP_001276506.1 | 8 - 9 | 1063 | 66 | |
NM_001289578.1 | NP_001276507.1 | 7 - 8 | 847 | 66 | ||
NM_001289579.1 | NP_001276508.1 | 9 - 10 | 1136 | 66 | ||
NM_001289580.1 | NP_001276509.1 | 9 - 10 | 1233 | 66 | ||
NM_172717.4 | NP_766305.2 | 8 - 9 | 1063 | 66 | ||
GenBank mRNA | AK077629.1 | - | 8 - 9 | 1035 | 66 | |
AK147892.1 | - | 7 - 8 | 822 | 66 | ||
AK148436.1 | - | 8 - 9 | 1034 | 66 | ||
AK154038.1 | - | 8 - 9 | 1045 | 66 | ||
AK169182.1 | - | 9 - 10 | 1089 | 66 | ||
AK169193.1 | - | 8 - 9 | 1018 | 66 | ||
BC049792.1 | - | 8 - 9 | 1017 | 6510480 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus Probe spans exons |