Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: P2rx2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164833.1 | NP_001158305.1 | 1 - 2 | 257 | 65 | |
NM_001164834.1 | NP_001158306.1 | 1 - 2 | 257 | 65 | ||
NM_153400.4 | NP_700449.2 | 1 - 2 | 257 | 65 | ||
GenBank mRNA | AB094663.1 | - | 1 - 2 | 177 | 65 | |
AB094664.1 | - | 1 - 2 | 177 | 65 | ||
AK141196.1 | - | 1 - 2 | 216 | 65 | ||
AY044240.2 | - | 1 - 2 | 433 | 65 | ||
BY236715.1 | - | 1 - 2 | 260 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Ion Channel Detect Genomic DNA Mus musculus 1700 Inventoried Probe spans exons |