Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Car11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009800.4 | NP_033930.1 | 2 - 3 | 447 | 68 | |
GenBank mRNA | AB079601.1 | - | 2 - 3 | 226 | 68 | |
AK013465.1 | - | 2 - 3 | 449 | 68 | ||
AK140073.1 | - | 2 - 3 | 548 | 68 | ||
AY075026.1 | - | 2 - 3 | 448 | 68 | ||
BC019393.1 | - | 2 - 3 | 432 | 3968862 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |