Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Htatip2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001146049.1 | NP_001139521.1 | 4 - 5 | 709 | 95 | |
NM_001146050.1 | NP_001139522.1 | 4 - 5 | 543 | 95 | ||
NM_001146052.1 | NP_001139524.1 | 3 - 4 | 533 | 95 | ||
NM_001146053.1 | NP_001139525.1 | 4 - 5 | 487 | 95 | ||
NM_016865.3 | NP_058561.2 | 4 - 5 | 653 | 95 | ||
GenBank mRNA | AF061972.1 | - | 3 - 4 | 637 | 95 | |
AK008630.1 | - | 3 - 4 | 528 | 95 | ||
AK144837.1 | - | 4 - 5 | 687 | 95 | ||
AK146400.1 | - | 3 - 4 | 348 | 95 | ||
AK151254.1 | - | 4 - 5 | 574 | 95 | ||
AK152418.1 | - | 4 - 5 | 574 | 95 | ||
BB575839.2 | - | 4 - 5 | 487 | 95 | ||
BC004083.1 | - | 4 - 5 | 606 | 3593077 | 95 | |
BC017372.1 | - | 3 - 4 | 499 | 4013057 | 95 | |
CB318388.1 | - | 3 - 4 | 536 | 95 | ||
CB593552.1 | - | 4 - 5 | 535 | 95 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Apoptosis Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons |