Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0700007744.mm.1 |
TC0700000870.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700007744.mm.1 |
NM_153057.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700007744.mm.1 |
NM_153057.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700007746.mm.1 |
TC0700000870.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700007746.mm.1 |
NM_153057.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700007746.mm.1 |
NM_153057.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700007743.mm.1 |
TC0700000870.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700007743.mm.1 |
NM_153057.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700007743.mm.1 |
NM_153057.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700007751.mm.1 |
TC0700000870.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700007751.mm.1 |
NM_153057.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700007751.mm.1 |
NM_153057.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700007741.mm.1 |
TC0700000870.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700007741.mm.1 |
NM_153057.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700007741.mm.1 |
NM_153057.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700007742.mm.1 |
TC0700000870.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700007742.mm.1 |
NM_153057.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700007742.mm.1 |
NM_153057.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700007748.mm.1 |
TC0700000870.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700007748.mm.1 |
NM_153057.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700007748.mm.1 |
NM_153057.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700007749.mm.1 |
TC0700000870.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700007749.mm.1 |
NM_153057.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700007749.mm.1 |
NM_153057.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700007745.mm.1 |
TC0700000870.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700007745.mm.1 |
NM_153057.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700007745.mm.1 |
NM_153057.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700007747.mm.1 |
TC0700000870.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700007747.mm.1 |
NM_153057.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700007747.mm.1 |
NM_153057.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700007740.mm.1 |
TC0700000870.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700007740.mm.1 |
NM_153057.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700007740.mm.1 |
NM_153057.4 |
Refseq |