Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sergef | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013789.2 | NP_038817.1 | 2 - 3 | 225 | 72 | |
GenBank mRNA | AJ243952.1 | - | 2 - 3 | 208 | 72 | |
AK145819.1 | - | 2 - 3 | 214 | 72 | ||
AK147848.1 | - | 2 - 3 | 219 | 72 | ||
BC049603.1 | - | 2 - 3 | 204 | 6591072 | 72 | |
BC119533.1 | - | 2 - 3 | 208 | 40056360 | 72 | |
BC119534.1 | - | 2 - 3 | 208 | 40056367 | 72 | |
BY228093.1 | - | 2 - 3 | 226 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |