Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dkkl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_015789.3 | NP_056604.2 | 1 - 2 | 130 | 58 | |
GenBank mRNA | AF177399.1 | - | 1 - 2 | 35 | 58 | |
AF274312.1 | - | 1 - 2 | 89 | 58 | ||
AK006175.1 | - | 1 - 2 | 397 | 58 | ||
AK131694.1 | - | 1 - 2 | 75 | 58 | ||
BC002215.1 | - | 1 - 2 | 108 | 3490482 | 58 | |
CA466481.1 | - | 1 - 2 | 133 | 58 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons |