Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Oas3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145226.2 | NP_660261.1 | 3 - 4 | 669 | 105 | |
GenBank mRNA | AB067534.1 | - | 3 - 4 | 669 | 105 | |
AF453830.1 | - | 3 - 4 | 105 | |||
AK158392.1 | - | 3 - 4 | 651 | 105 | ||
BC141055.1 | - | 3 - 4 | 662 | 9055667 | 105 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |