Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Iqcd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029408.2 | NP_083684.1 | 3 - 4 | 1105 | 84 | |
GenBank mRNA | AK017032.1 | - | 3 - 4 | 1105 | 84 | |
BC125429.1 | - | 3 - 4 | 1099 | 40129944 | 84 | |
BC137644.1 | - | 3 - 4 | 1099 | 8860663 | 84 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |