Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdyhv1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029734.1 | NP_084010.1 | 3 - 4 | 305 | 80 | |
GenBank mRNA | AK010831.1 | - | 3 - 4 | 326 | 80 | |
AK150578.1 | - | 3 - 4 | 357 | 80 | ||
BC051524.1 | - | 3 - 4 | 305 | 1514917 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |