Array Name |
Probe IDs |
Targets |
Target Type |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2123593 |
NM_001104627.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P1970887 |
NM_001104627.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P1970887 |
NM_001104625.1 |
mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
635270 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
442521 |
AK021089.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
442521 |
AK039322.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
442521 |
AK122252.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
442521 |
AY528242.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
442521 |
BC055053.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
143434 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
734644 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
497706 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
124488 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
856926 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
323576 |
AF032459.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
323576 |
AF032460.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
323576 |
AF032461.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
323576 |
AK011490.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
323576 |
AK050192.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
323576 |
AK082613.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
323576 |
AK150464.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
323576 |
AK152472.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
323576 |
AK153542.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
323576 |
BC019556.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
323576 |
NM_009754.3 |
refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
323576 |
NM_207680.2 |
refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
323576 |
NM_207681.2 |
refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
468343 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
283581 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
158065 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
32488 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
58461 |
- |
- |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2123593 |
NM_001104627.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P1970887 |
NM_001104627.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P1970887 |
NM_001104625.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P4296 |
NM_001104620.1 |
mouse_transcriptome |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23975779 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23975794 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23975760 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23975781 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23975730 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23975769 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23975776 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23975722 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23975777 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23975724 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23975743 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23975762 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23975801 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23975770 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23975742 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23975723 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23975740 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23975737 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23975780 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23975725 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23975717 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23975712 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23975718 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23975741 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23975775 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23975715 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23975783 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23975717 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23975715 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23975728 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23975732 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23975723 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23975721 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23975730 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23975736 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23975718 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23975722 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23975734 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23975737 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23975725 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23975727 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P04969069 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23975724 |
- |
- |