Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Paqr3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198422.2 | NP_940814.1 | 1 - 2 | 344 | 71 | |
GenBank mRNA | AK149213.1 | - | 1 - 2 | 323 | 71 | |
AK158711.1 | - | 1 - 2 | 332 | 71 | ||
AK172689.1 | - | 1 - 2 | 299 | 71 | ||
AY424292.1 | - | 1 - 2 | 344 | 71 | ||
BC050248.1 | - | 1 - 2 | 344 | 71 | ||
BC051173.1 | - | 1 - 2 | 344 | 71 | ||
BC120853.1 | - | 1 - 2 | 329 | 40129776 | 71 | |
BC120855.1 | - | 1 - 2 | 330 | 40129778 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |