Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zkscan1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029869.1 | NP_084145.1 | 2 - 3 | 653 | 107 | |
NM_133906.4 | NP_598667.2 | 2 - 3 | 659 | 107 | ||
GenBank mRNA | AK020039.1 | - | 2 - 3 | 635 | 107 | |
AK028932.1 | - | 2 - 3 | 590 | 107 | ||
AK031759.1 | - | 2 - 3 | 640 | 107 | ||
AK036807.1 | - | 2 - 3 | 642 | 107 | ||
AK054323.1 | - | 2 - 3 | 630 | 107 | ||
AK131148.1 | - | 1 - 2 | 798 | 107 | ||
AK135999.1 | - | 2 - 3 | 609 | 107 | ||
AK154831.1 | - | 2 - 3 | 639 | 107 | ||
BC052441.1 | - | 2 - 3 | 617 | 6406518 | 107 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus Inventoried Probe spans exons |