Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pxn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011223.3 | NP_035353.1 | 6 - 7 | 1017 | 60 | |
GenBank mRNA | AF293882.1 | - | 6 - 7 | 942 | 60 | |
AK148060.1 | - | 6 - 7 | 1003 | 60 | ||
AK149933.1 | - | 6 - 7 | 982 | 60 | ||
AK167299.1 | - | 6 - 7 | 956 | 60 | ||
AK171226.1 | - | 6 - 7 | 1005 | 60 | ||
AK171436.1 | - | 6 - 7 | 982 | 60 | ||
BC003298.1 | - | 2 - 3 | 300 | 60 | ||
BC025493.1 | - | 6 - 7 | 975 | 60 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
---|---|---|---|---|---|---|---|
GenBankmRNA | AK089534.1 | - | Chr. - - [-] on Build | 60 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |