Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ankrd13a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026718.2 | NP_080994.2 | 1 - 2 | 184 | 91 | |
GenBank mRNA | AK039432.1 | - | 1 - 2 | 377 | 91 | |
AK156539.1 | - | 1 - 2 | 377 | 91 | ||
BC049187.1 | - | 1 - 2 | 363 | 91 | ||
BC061692.1 | - | 1 - 2 | 184 | 6308996 | 91 | |
BC108413.1 | - | 1 - 2 | 659 | 6487251 | 91 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |