Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ddx54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028041.2 | NP_082317.1 | 4 - 5 | 570 | 69 | |
GenBank mRNA | AF319547.1 | - | 4 - 5 | 930 | 69 | |
AK028274.1 | - | 4 - 5 | 584 | 69 | ||
AK031534.1 | - | 4 - 5 | 580 | 69 | ||
AK147715.1 | - | 4 - 5 | 585 | 69 | ||
AK155138.1 | - | 4 - 5 | 582 | 69 | ||
AK167603.1 | - | 4 - 5 | 584 | 69 | ||
AK168455.1 | - | 4 - 5 | 584 | 69 | ||
AK169255.1 | - | 4 - 5 | 583 | 69 | ||
AK170967.1 | - | 4 - 5 | 582 | 69 | ||
BC043699.1 | - | 4 - 5 | 570 | 5344158 | 69 | |
BC062920.1 | - | 4 - 5 | 573 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |